Merck
  • In vivo induction of CYP in mice by carbamazepine is independent on PXR.
In vivo induction of CYP in mice by carbamazepine is independent on PXR.
Pharmacological reports : PR (2015-02-26)
Aiming Liu, Chuang Wang, Michael Hehir, Tianbao Zhou, Julin Yang
ABSTRACT
The antiepileptic drug carbamazepine (CBZ) is a typical inducer of cytochrome P450 (CYP) 3A and 2C in the clinic. It is considered a strong constitutive androstane receptor activator, however both CBZ and its main metabolite CBZ 10, 11-epoxide have been reported to be pregnane X receptor (PXR) activators whose maximal efficacy and potency are comparable with the human PXR ligand rifampicin. It is unknown whether or not PXR plays a substantially important role in in vivo induction of CYP by CBZ administration. In this study, wild type and Pxr-/- mice were administered with CBZ for 5 days. Serum and liver samples were collected and subjected to hepatotoxicity assessment and CYP induction analysis. CYP2b, 2c and 3a were induced similarly in terms of transcription level, enzyme activity and protein abundance in both wild type and Pxr-/- mice. Inductive profile of CYPs in mice by CBZ administration accorded with those reported in rats, but differed from clinically reported data. These data suggest that in vivo induction of CYP in mice by multiple administration of CBZ is independent of PXR. Knowledge of the featured CYP induction profile in mice helps us understand species related CYP induction profiles among rodents and humans resulting from administration of CBZ.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Supelco
Chlorpropamide, analytical standard, ≥97%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®