Merck
  • Antitumor activity of the MEK inhibitor TAK-733 against melanoma cell lines and patient-derived tumor explants.
Antitumor activity of the MEK inhibitor TAK-733 against melanoma cell lines and patient-derived tumor explants.
Molecular cancer therapeutics (2014-11-08)
Lindsey N Micel, John J Tentler, Aik-Choon Tan, Heather M Selby, Kelsey L Brunkow, Kelli M Robertson, S Lindsey Davis, Peter J Klauck, Todd M Pitts, Esha Gangolli, Robyn Fabrey, Shawn M O'Connell, Patrick W Vincent, S Gail Eckhardt
ABSTRACT
The goal of this study was to investigate the activity of the selective MEK1/2 inhibitor TAK-733 in both melanoma cell lines and patient-derived melanoma xenograft models. In vitro cell proliferation assays using the sulforhodamine B assay were conducted to determine TAK-733 potency and melanoma responsiveness. In vivo murine modeling with eleven patient-derived melanoma explants evaluated daily dosing of TAK-733 at 25 or 10 mg/kg. Immunoblotting was performed to evaluate on-target activity and downstream inhibition by TAK-733 in both in vitro and in vivo studies. TAK-733 demonstrated broad activity in most melanoma cell lines with relative resistance observed at IC50 > 0.1 μmol/L in vitro. TAK-733 also exhibited activity in 10 out of 11 patient-derived explants with tumor growth inhibition ranging from 0% to 100% (P < 0.001-0.03). Interestingly, BRAF(V600E) and NRAS mutational status did not correlate with responsiveness to TAK-733. Pharmacodynamically, pERK was suppressed in sensitive cell lines and tumor explants, confirming TAK-733-mediated inhibition of MEK1/2, although the demonstration of similar effects in the relatively resistant cell lines and tumor explants suggests that escape pathways are contributing to melanoma survival and proliferation. These data demonstrate that TAK-733 exhibits robust tumor growth inhibition and regression against human melanoma cell lines and patient-derived xenograft models, suggesting that further clinical development in melanoma is of scientific interest. Particularly interesting is the activity in BRAF wild-type models, where current approved therapy such as vemurafenib has been reported not to be active.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethanesulfonic acid, 95%
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%