Merck
  • Characterization of drug-resistant influenza virus A(H1N1) and A(H3N2) variants selected in vitro with laninamivir.
Characterization of drug-resistant influenza virus A(H1N1) and A(H3N2) variants selected in vitro with laninamivir.
Antimicrobial agents and chemotherapy (2014-06-25)
Mélanie Samson, Yacine Abed, François-Marc Desrochers, Stephanie Hamilton, Angela Luttick, Simon P Tucker, Melinda J Pryor, Guy Boivin
ABSTRACT
Neuraminidase inhibitors (NAIs) play a major role for managing influenza virus infections. The widespread oseltamivir resistance among 2007-2008 seasonal A(H1N1) viruses and community outbreaks of oseltamivir-resistant A(H1N1)pdm09 strains highlights the need for additional anti-influenza virus agents. Laninamivir is a novel long-lasting NAI that has demonstrated in vitro activity against influenza A and B viruses, and its prodrug (laninamivir octanoate) is in phase II clinical trials in the United States and other countries. Currently, little information is available on the mechanisms of resistance to laninamivir. In this study, we first performed neuraminidase (NA) inhibition assays to determine the activity of laninamivir against a set of influenza A viruses containing NA mutations conferring resistance to one or many other NAIs. We also generated drug-resistant A(H1N1) and A(H3N2) viruses under in vitro laninamivir pressure. Laninamivir demonstrated a profile of susceptibility that was similar to that of zanamivir. More specifically, it retained activity against oseltamivir-resistant H275Y and N295S A(H1N1) variants and the E119V A(H3N2) variant. In vitro, laninamivir pressure selected the E119A NA substitution in the A/Solomon Islands/3/2006 A(H1N1) background, whereas E119K and G147E NA changes along with a K133E hemagglutinin (HA) substitution were selected in the A/Quebec/144147/2009 A(H1N1)pdm09 strain. In the A/Brisbane/10/2007 A(H3N2) background, a large NA deletion accompanied by S138A/P194L HA substitutions was selected. This H3N2 variant had altered receptor-binding properties and was highly resistant to laninamivir in plaque reduction assays. Overall, we confirmed the similarity between zanamivir and laninamivir susceptibility profiles and demonstrated that both NA and HA changes can contribute to laninamivir resistance in vitro.
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zanamivir, ≥98% (HPLC)
USP
Zanamivir, United States Pharmacopeia (USP) Reference Standard