Merck
  • Plasma folate, vitamin B12, and homocysteine and cancers of the esophagus, stomach, and liver in a Chinese population.
Plasma folate, vitamin B12, and homocysteine and cancers of the esophagus, stomach, and liver in a Chinese population.
Nutrition and cancer (2015-01-22)
Shen-Chih Chang, Binh Y Goldstein, Lina Mu, Lin Cai, Nai-Chieh Y You, Na He, Bao-Guo Ding, Jin-Kou Zhao, Shun-Zhang Yu, David Heber, Zuo-Feng Zhang, Qing-Yi Lu
ABSTRACT
Evidence is accumulating regarding a role of micronutrients in folate metabolism in cancer risk. We investigated the associations of plasma folate, vitamin B12, and homocysteine with upper gastrointestinal (GI) cancers in a population-based case-control study in Taixing City, China. With informed consent, we recruited cases with cancers of esophagus (n = 218), stomach (n = 206), and liver (n = 204), and one common healthy control group (n = 405). A standardized epidemiologic questionnaire was used in face-to-face interviews, and blood samples were collected during interviews. We observed an inverse association between plasma folate levels and liver cancer. The adjusted odds ratio (aOR) was 0.46 [95% confidence interval (CI) = 0.24-0.88] comparing individuals in the highest quartile to those in the lowest. We found a positive association between plasma vitamin B12 levels and all three cancers. The aORs for those in the highest quartile were 2.80 (95% CI = 1.51-5.18) for esophageal cancer, 2.17 (1.21-3.89) for stomach cancer, and 9.97 (4.82-20.60) for liver cancer, comparing to those in the lowest quartile. We further observed interaction between plasma folate and vitamin B12 on these cancers. Our data indicated associations between plasma folate and vitamin B12 with upper GI cancers in Chinese population. Further research is warranted considering the debate over the necessity of food fortification.
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Supelco
Cyanocobalamin (B12), analytical standard
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%