Merck
  • Effect of acid-catalyzed formation rates of benzimidazole-linked polymers on porosity and selective CO2 capture from gas mixtures.
Effect of acid-catalyzed formation rates of benzimidazole-linked polymers on porosity and selective CO2 capture from gas mixtures.
Environmental science & technology (2015-03-03)
Suha Altarawneh, Timur İslamoğlu, Ali Kemal Sekizkardes, Hani M El-Kaderi
ABSTRACT
Benzimidazole-linked polymers (BILPs) are emerging candidates for gas storage and separation applications; however, their current synthetic methods offer limited control over textural properties which are vital for their multifaceted use. In this study, we investigate the impact of acid-catalyzed formation rates of the imidazole units on the porosity levels of BILPs and subsequent effects on CO2 and CH4 binding affinities and selective uptake of CO2 over CH4 and N2. Treatment of 3,3'-Diaminobenzidine tetrahydrochloride hydrate with 1,2,4,5-tetrakis(4-formylphenyl)benzene and 1,3,5-(4-formylphenyl)-benzene in anhydrous DMF afforded porous BILP-15 (448 m(2) g(-1)) and BILP-16 (435 m(2) g(-1)), respectively. Alternatively, the same polymers were prepared from the neutral 3,3'-Diaminobenzidine and catalytic amounts of aqueous HCl. The resulting polymers denoted BILP-15(AC) and BILP-16(AC) exhibited optimal surface areas; 862 m(2) g(-1) and 643 m(2) g(-1), respectively, only when 2 equiv of HCl (0.22 M) was used. In contrast, the CO2 binding affinity (Qst) dropped from 33.0 to 28.9 kJ mol(-1) for BILP-15 and from 32.0 to 31.6 kJ mol(-1) for BILP-16. According to initial slope calculations at 273 K/298 K, a notable change in CO2/N2 selectivity was observed for BILP-15(AC) (61/50) compared to BILP-15 (83/63). Similarly, ideal adsorbed solution theory (IAST) calculations also show the higher specific surface area of BILP-15(AC) and BILP-16(AC) compromises their CO2/N2 selectivity.
Product Description
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,4-Dioxane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
1,4-Dioxane, analytical standard
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
1,4-Dioxane, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
2-Methyl-2-butene, ≥95.0% (GC)
Supelco
2-Methyl-2-butene, analytical standard
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Supelco
Ethyl acetate, analytical standard