Merck
  • Coordinated expression of H3K9 histone methyltransferases during tooth development in mice.
Coordinated expression of H3K9 histone methyltransferases during tooth development in mice.
Histochemistry and cell biology (2014-10-09)
Taichi Kamiunten, Hisashi Ideno, Akemi Shimada, Yoshiki Nakamura, Hiroshi Kimura, Kazuhisa Nakashima, Akira Nifuji
ABSTRACT
Tissue-specific gene expression is subjected to epigenetic and genetic regulation. Posttranslational modifications of histone tails alter the accessibility of nuclear proteins to DNA, thus affecting the activity of the regulatory complex of nuclear proteins. Methylation at histone 3 lysine 9 (H3K9) is a crucial modification that affects gene expression and cell differentiation. H3K9 is known to have 0-3 methylation states, and these four methylated states are determined by the expression of sets of histone methyltransferases. During development, teeth are formed through mutual interactions between the mesenchyme and epithelium via a process that is subjected to the epigenetic regulation. In this study, we examined the expression of all H3K9 methyltransferases (H3K9MTases) during mouse tooth development. We found that four H3K9MTases-G9a, Glp, Prdm2, and Suv39h1-were highly expressed in the tooth germ, with expression peaks at around embryonic days 16.5 and 17.5 in mice. Immunohistochemical and in situ hybridization analyses revealed that all four H3K9MTases were enriched in the mesenchyme more than in the epithelium. Substrates of H3K9MTases, H3K9me1, H3K9me2, and H3K9me3 were also enriched in the mesenchyme. Taken together, these data suggested that coordinated expression of four H3K9MTases in the dental mesenchyme might play important roles in tooth development.
Product Description
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Chloroform, analytical standard
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material