Merck
  • Efficacy and safety of canagliflozin alone or as add-on to other oral antihyperglycemic drugs in Japanese patients with type 2 diabetes: A 52-week open-label study.
Efficacy and safety of canagliflozin alone or as add-on to other oral antihyperglycemic drugs in Japanese patients with type 2 diabetes: A 52-week open-label study.
Journal of diabetes investigation (2015-03-25)
Nobuya Inagaki, Kazuoki Kondo, Toru Yoshinari, Hideki Kuki
ABSTRACT
Canagliflozin is a sodium-glucose cotransporter 2 inhibitor under development for the treatment of type 2 diabetes. Our aim was to examine its efficacy and safety as monotherapy or in combination with commonly used oral antihyperglycemic drugs in Japanese patients with type 2 diabetes. Patients on diet/exercise alone or diet/exercise plus an oral antihyperglycemic drug (sulfonylurea, glinide, α-glucosidase inhibitor, biguanide, thiazolidinedione or dipeptidyl peptidase-4 inhibitor) were randomized to either 100 or 200 mg canagliflozin while continuing prior therapy. Patients were treated for 52 weeks in an open-label manner. Canagliflozin significantly reduced hemoglobin A1c, fasting plasma glucose and bodyweight in all the study groups. Improvements were apparent by 4 weeks of treatment, and were maintained for 52 weeks. The reduction in hemoglobin A1c ranged from -0.80 to -1.06%, and from -0.93 to -1.26% in the 100 and 200 mg canagliflozin groups, respectively. Drug-related adverse events occurred in approximately one-third of patients, and included hypoglycemia/asymptomatic hypoglycemia and pollakiuria. Hypoglycemia/asymptomatic hypoglycemia was most common in patients treated with a sulfonylurea. Most adverse events were classified as mild or moderate in severity. The results of the present study confirmed that treatment with canagliflozin resulted in significant reductions in glycemic control and bodyweight that were maintained for 52 weeks of treatment irrespective of whether it was administered as monotherapy or in combination with another oral antihyperglycemic drug. Canagliflozin was well tolerated, with a low incidence of drug-related adverse events. This trial was registered with ClinicalTrials.gov (no. NCT01387737).
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard