Merck
  • Mechanistic Insight into the Yolk@Shell Transformation of MnO@Silica Nanospheres Incorporating Ni(2+) Ions toward a Colloidal Hollow Nanoreactor.
Mechanistic Insight into the Yolk@Shell Transformation of MnO@Silica Nanospheres Incorporating Ni(2+) Ions toward a Colloidal Hollow Nanoreactor.
Small (Weinheim an der Bergstrasse, Germany) (2014-12-17)
Jin Goo Kim, Soo Min Kim, In Su Lee
ABSTRACT
As an effort to develop a simple and versatile synthetic strategy that contributes to the evolution of hollow nanostructures with increasing complexity and functionality, this research is devoted to study the hollow transformation within a nanosized solid matrix. Through an in-depth investigation of a hollowing process of MnO nanocrystals confined within a Ni(2+) incorporating silica nanosphere, a very distinct transformation pathway can be explained that produces the yolk@shell nanostructure with a single Ni nanocrystal inside a silicate nanoshell. The yolk@shell structure is developed by a mechanism combining different processes, including the formation of a (Ni0.1 Mn0.9 )O mixed-metal oxide and subsequent segregation of the reduced Ni. Furthermore, this study also devises a protocol to exploit the solid-state-synthesized powder for fabricating a colloidal hollow nanoreactor that can selectively catalyze the reduction of nitroarenes and be recycled via the magnetic process.
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
4-Nitroanisole, 97%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium oleate, ≥95% (capillary GC)
Sigma-Aldrich
Sodium oleate, ≥99%
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
4-Nitroaniline, ≥99%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium oleate, ≥82% (fatty acids), powder
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Supelco
1-Octadecene, analytical standard, ≥99.0% (GC)
Supelco
4-Nitroaniline, analytical standard
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
2-Amino-4-nitrophenol, 96%
Sigma-Aldrich
2-Nitrophenol, 98%
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Nitrobenzaldehyde, 98% (GC)
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
VenPure® SF, powder
Supelco
4-Nitrophenol, PESTANAL®, analytical standard