Merck
  • Radiation exposure induces inflammasome pathway activation in immune cells.
Radiation exposure induces inflammasome pathway activation in immune cells.
Journal of immunology (Baltimore, Md. : 1950) (2014-12-30)
Veit M Stoecklein, Akinori Osuka, Shizu Ishikawa, Madeline R Lederer, Lorenz Wanke-Jellinek, James A Lederer
ABSTRACT
Radiation exposure induces cell and tissue damage, causing local and systemic inflammatory responses. Because the inflammasome pathway is triggered by cell death and danger-associated molecular patterns, we hypothesized that the inflammasome may signal acute and chronic immune responses to radiation. Using a mouse radiation model, we show that radiation induces a dose-dependent increase in inflammasome activation in macrophages, dendritic cells, NK cells, T cells, and B cells as judged by cleaved caspase-1 detection in cells. Time course analysis showed the appearance of cleaved caspase-1 in cells by day 1 and sustained expression until day 7 after radiation. Also, cells showing inflammasome activation coexpressed the cell surface apoptosis marker annexin V. The role of caspase-1 as a trigger for hematopoietic cell losses after radiation was studied in caspase-1(-/-) mice. We found less radiation-induced cell apoptosis and immune cell loss in caspase-1(-/-) mice than in control mice. Next, we tested whether uric acid might mediate inflammasome activation in cells by treating mice with allopurinol and discovered that allopurinol treatment completely blocked caspase-1 activation in cells. Finally, we demonstrate that radiation-induced caspase-1 activation occurs by a Nod-like receptor family protein 3-independent mechanism because radiation-exposed Nlrp3(-/-) mice showed caspase-1 activation profiles that were indistinguishable from those of wild-type mice. In summary, our data demonstrate that inflammasome activation occurs in many immune cell types following radiation exposure and that allopurinol prevented radiation-induced inflammasome activation. These results suggest that targeting the inflammasome may help control radiation-induced inflammation.
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Allopurinol, European Pharmacopoeia (EP) Reference Standard
Supelco
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Allopurinol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)