Merck
  • A sensitive method for digoxin determination using formate-adduct ion based on the effect of ionization enhancement in liquid chromatograph-mass spectrometer.
A sensitive method for digoxin determination using formate-adduct ion based on the effect of ionization enhancement in liquid chromatograph-mass spectrometer.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-30)
Xia Li, Yu Wang, Qingyuan Zhou, Yunqiu Yu, Lihong Chen, Jie Zheng
ABSTRACT
A sensitive and rapid method based on formate-adduct ion detection was developed and fully validated for digoxin determination in rat plasma. For LC/MS/MS detection with formate-adducts as precursor ions, transitions of m/z 825.5→779.9 for digoxin and m/z 809.5→763.4 for the internal standard (digitoxin) were monitored in negative mode. To investigate the impact of formic acid on the mass response and method sensitivity, a formic acid concentration range of 0-0.1% (0, 0.0005%, 0.002%, 0.01%, 0.1%, v/v) was evaluated. A concentration of 0.002% gave the highest sensitivity, which was 16- to 18-fold higher than deprotonated ions, and was designated as the contribution giving the strongest ionization enhancement and adduction. A number of parameters were then varied in order to optimize the method, and a limit of quantitation (LOQ) at 0.2 ng/mL was reached with an injection volume of 5 μL, a total run time of 3 min, and 0.1 mL of rat plasma. A calibration curve was plotted over the range 0.2-50 ng/mL (R(2)=0.9998), and the method was successfully applied to study pharmacokinetics in rat following a single oral administration of digoxin (0.05 mg/kg). Four additional steroid saponins (digitoxin, deslanoside, ginsenoside Rg1 and Rb1) were investigated to assess the impact of formic acid on the mass response of steroid saponins. Compounds with a conjugated lactonic ring in their structures such as digoxin, digitoxin and deslanoside tended to form stable formate-adduct ions more easily. The LC/MS/MS method developed here is therefore well suited for the quantification of steroid saponins that are difficult to deprotonate using other MS approaches.
Product Description
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Digoxin, analytical standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Digoxin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Digoxin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ultrapure Acetonitrile
Digoxin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard