Merck
  • Aptamer-functionalized superporous hydrogels for sequestration and release of growth factors regulated via molecular recognition.
Aptamer-functionalized superporous hydrogels for sequestration and release of growth factors regulated via molecular recognition.
Biomaterials (2014-06-24)
Mark R Battig, Yike Huang, Niancao Chen, Yong Wang
ABSTRACT
While the discovery of highly potent biologics has led to the development of promising therapies for various human diseases, biologics can cause severe toxicity if delivered inappropriately. Thus, great efforts have been made to synthesize polymeric systems for safe and efficient delivery of biologics. However, the application of polymeric delivery systems is often limited by problems such as harsh reaction conditions, low drug sequestration efficiency, and difficult drug release regulation. This study was aimed at developing a superporous material system with a hydrogel and an aptamer to overcome these challenges. The results have shown that the superporous hydrogel is capable of instantaneously and fully sequestering a large amount of growth factors, owing to the presence of superporous architectures and aptamers. Moreover, the sequestering and loading procedure does not involve any harsh conditions. The release kinetics of growth factors can be molecularly modulated by either changing the binding affinity of the aptamer or by using a triggering effector. Therefore, this study presents a promising superporous material for the delivery of highly potent biologics such as growth factors for clinical applications.
Product Description
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
Acetic acid, analytical standard