Merck
  • Cytotoxic and apoptotic activities of the plancitoxin I from the venom of crown-of-thorns starfish (Acanthaster planci) on A375.S2 cells.
Cytotoxic and apoptotic activities of the plancitoxin I from the venom of crown-of-thorns starfish (Acanthaster planci) on A375.S2 cells.
Journal of applied toxicology : JAT (2014-07-23)
Chi-Chiu Lee, Hernyi Justin Hsieh, Deng-Fwu Hwang
ABSTRACT
This study reports on a cytotoxic toxin derived from the venom of the crown-of-thorns starfish Acanthaster planci (CAV). The protein toxin was isolated through both ion-exchange and gel-filtration chromatography, and characterized by sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) and mass spectrum analyzes. The CAV was identified as plancitoxin I protein. The mechanistic role of the CAV toxin was explored in human malignant melanoma A375.S2 cell death. The results indicated that after incubation with CAV toxin, cells significantly decreased in A375.S2 cell viability and increased in the lactate dehydrogenase (LDH) level in a dose-dependent manner. The assays indicated that CAV toxin promoted reactive oxygen species (ROS) production, induced nitric oxide (NO) formation, lost mitochondrial membrane potential (ΔΨm) and induced inter-nucleosomal DNA fragmentation in A375.S2 cells. The molecular cytotoxicity of the CAV toxin was tested through evaluation of the apoptosis/necrosis ratio by double staining with annexin V-FITC and a propidium iodide (PI) assay. The results suggested that CAV toxin induced a cytotoxic effect in A375.S2 cells via the apoptotic procedure, and may be associated with the regulation of the p38 pathways.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Supelco
2-Butanone, analytical standard
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
Rhodamine 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Methyl Ethyl Ketone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
USP
Sulfanilamide, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sulfanilamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis