Merck
  • Bioresponsive nanohydrogels based on HEAA and NIPA for poorly soluble drugs delivery.
Bioresponsive nanohydrogels based on HEAA and NIPA for poorly soluble drugs delivery.
International journal of pharmaceutics (2014-05-13)
Elena Pérez, Ana Martínez, César Teijón, Jose M Teijón, M Dolores Blanco
ABSTRACT
Environmentally sensitive hydrogels have gained considerable attention in recent years as one of the most promising drug delivery systems. In the present study, two new formulations of pH and temperature stimuli-responsive nanogels (NGs) based on poly-N-isopropylacrylamide (NIPA), N-hydroxyethyl acrylamide (HEAA) and tert-butyl 2-acrylamidoethyl carbamate (2AAECM) were synthesized and evaluated for passive targeting of paclitaxel (PTX). Nanogels were prepared by microemulsion polymerization method using N-methylenebis(acrylamide) (NMBA) as crosslinking agent. TEM images and DLS results showed nanosized spherical hydrogels. FTIR spectra confirmed the synthesis of nanogels by radical polymerization among vinyl groups of monomers. The PTX loading capacity, encapsulation efficiency and in vitro release were analyzed by HPLC. The cumulative release profile of the PTX-loaded nanohydrogels within 144h showed a faster drug release at acid pH (pH 5), similar to those observed at lysosome compartment, whereas a fewer PTX amount was released from NGs at pH similar to plasma levels. Cellular uptake assays revealed rapid penetration and intracellular accumulation of those nanogels in MCF7, HeLa and T47D cells after 48h incubation. MTT assays showed cell viability dependence on concentration and time incubation. Finally, the PTX effect on cell viability showed a G2/M cell arrest after using PTX-loaded NGs and pure PTX.
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 1.0 M in THF
Supelco
Ethylenediamine, analytical standard
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in methylene chloride
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in THF
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Citric acid, 99%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland