Merck
  • Nitrite dismutase reaction mechanism: kinetic and spectroscopic investigation of the interaction between nitrophorin and nitrite.
Nitrite dismutase reaction mechanism: kinetic and spectroscopic investigation of the interaction between nitrophorin and nitrite.
Journal of the American Chemical Society (2015-03-10)
Chunmao He, Barry D Howes, Giulietta Smulevich, Sigrun Rumpel, Edward J Reijerse, Wolfgang Lubitz, Nicholas Cox, Markus Knipp
ABSTRACT
Nitrite is an important metabolite in the physiological pathways of NO and other nitrogen oxides in both enzymatic and nonenzymatic reactions. The ferric heme b protein nitrophorin 4 (NP4) is capable of catalyzing nitrite disproportionation at neutral pH, producing NO. Here we attempt to resolve its disproportionation mechanism. Isothermal titration calorimetry of a gallium(III) derivative of NP4 demonstrates that the heme iron coordinates the first substrate nitrite. Contrary to previous low-temperature EPR measurements, which assigned the NP4-nitrite complex electronic configuration solely to a low-spin (S = 1/2) species, electronic absorption and resonance Raman spectroscopy presented here demonstrate that the NP4-NO2(-) cofactor exists in a high-spin/low-spin equilibrium of 7:3 which is in fast exchange in solution. Spin-state interchange is taken as evidence for dynamic NO2(-) coordination, with the high-spin configuration (S = 5/2) representing the reactive species. Subsequent kinetic measurements reveal that the dismutation reaction proceeds in two discrete steps and identify an {FeNO}(7) intermediate species. The first reaction step, generating the {FeNO}(7) intermediate, represents an oxygen atom transfer from the iron bound nitrite to a second nitrite molecule in the protein pocket. In the second step this intermediate reduces a third nitrite substrate yielding two NO molecules. A nearby aspartic acid residue side-chain transiently stores protons required for the reaction, which is crucial for NPs' function as nitrite dismutase.
Sigma-Aldrich
N,N-Diethylaniline, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Nitrite ion standard solution, for ion-selective electrodes, 0.1 M NO2-
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium nitrite, analytical standard
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%