Merck
  • Effects of uremic solutes on reactive oxygen species in vitro model systems as a possibility of support the renal function management.
Effects of uremic solutes on reactive oxygen species in vitro model systems as a possibility of support the renal function management.
BMC nephrology (2015-04-18)
Renata P Assis, Juliana F A Castro, Vânia O Gutierres, Carlos A Arcaro, Renata S Brotto, Olga M M F Oliveira, Amanda M Baviera, Iguatemy L Brunetti
ABSTRACT
In view of the prevalence of oxidative stress in chronic kidney disease (CKD) patients, the loss of low-molecular-weight biomolecules by hemodialysis and the antioxidant potential of some uremic solutes that accumulate in CKD, we used in vitro model systems to test the antioxidant potential of the following uremic solutes: uric acid, hippuric acid, p-cresol, phenol, methylguanidine, L-arginine, L-tyrosine, creatinine and urea. The in vitro antioxidant efficiencies of the uremic solutes, isolated or in mixtures, were tested with the following assays: i) ABTS radical cation decolorization assay; ii) hypochlorous acid (HOCl/OCl(-)) scavenging activity; iii) superoxide anion radical (O2(•-)) scavenging activity; iv) crocin bleaching assay (capture of peroxyl radical, ROO(•)); v) hydrogen peroxide (H2O2) scavenging activity. Four of the tested uremic solutes (p-cresol, phenol, L-tyrosine, uric acid) were effective antioxidants and their IC50 were found in three model systems: ABTS(•+), HOCl/OCl(-) and crocin bleaching assay. In the 4-solutes mixtures, each one of the solute captured 12.5% for the IC50 of the mixture to ABTS(•+) or HOCl/OCl(-), exhibiting a virtually exact additive effect. In the 2-solutes mixtures, for ROO(•) capture, it was observed the need of more mass of uremic solutes to reach an IC50 value that was higher than the projected IC50, obtained from the IC50 of single solutes (25% of each, in the binary mixtures) in the same assay. In model systems for O2(•-) and H2O2, none of the uremic solutes showed scavenging activity. The use of the IC50 as an analytical tool to prepare and analyze mixtures allows the determination of their scavenging capacities and may be useful for the assessment of the antioxidant status of biological samples under conditions of altered levels of the endogenous antioxidant network and/or in the employment and monitoring of exogenous antioxidant therapy.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hippuric acid, 98%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
p-Cresol, analytical standard