Merck
  • Tumor-targeting and pH-sensitive lipoprotein-mimic nanocarrier for targeted intracellular delivery of paclitaxel.
Tumor-targeting and pH-sensitive lipoprotein-mimic nanocarrier for targeted intracellular delivery of paclitaxel.
International journal of pharmaceutics (2015-01-24)
Conghui Chen, Haiyang Hu, Mingxi Qiao, Xiuli Zhao, Yinjie Wang, Kang Chen, Xiong Guo, Dawei Chen
ABSTRACT
In the present study, we constructed a tumor-targeting and pH-sensitive lipoprotein-mimic nanocarrier containing paclitaxel (FA-BSA-LC/DOPE-PTX), by adding 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE) and oleic acid as pH-sensitive components into the formulation of lipid core and then coating with folic acid modified bovine serum albumin (FA-BSA) for tumor targeting activity. In vitro drug release study demonstrated that paclitaxel (PTX) was released from FA-BSA-LC/DOPE in a pH-dependent manner. The vitro cytotoxicity assays showed that all the blank nanocarriers were nontoxic. However, MTT assay showed that FA-BSA-LC/DOPE-PTX was highly cytotoxic. Cellular uptake experiments analyzed with flow cytometry and laser scan confocal microscope (LSCM) revealed that FA-BSA-LC/DOPE was taken up in great amount via folate receptor-mediated endocytosis and pH-sensitive release of drug to cytoplasm. Furthermore, the study of intracellular drug release behavior demonstrated that the FA-BSA-LC/DOPE escaped from lysosomes and released drug into cytoplasm. The in vivo targeting activity showed that the nanocarrier selectively targeted tumor and had long residence time for BSA layer increased the stability in blood. Moreover, FA-BSA-LC/DOPE-PTX produced very marked anti-tumor activity in tumor-bearing mice in vivo. Therefore, FA-BSA-LC/DOPE as biocompatible, tumor-targeting and pH-sensitive lipoprotein-mimic nanocarrier is a promising system for effective intracellular delivery of PTX to tumor.
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Octadecylamine, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Octadecylamine, technical grade, 90%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer