Merck
  • 3,6-Epidioxy-1,10-bisaboladiene inhibits G1 -specific transcription through Swi4/Swi6 and Mbp1/Swi6 via the Hog1 stress pathway in yeast.
3,6-Epidioxy-1,10-bisaboladiene inhibits G1 -specific transcription through Swi4/Swi6 and Mbp1/Swi6 via the Hog1 stress pathway in yeast.
The FEBS journal (2014-08-13)
Yuko Imamura, Masashi Yukawa, Masaru Ueno, Ken-Ichi Kimura, Eiko Tsuchiya
ABSTRACT
3,6-Epidioxy-1,10-bisaboladiene (EDBD), a bisabolane sesquiterpene endoperoxide compound, was previously isolated from Cacalia delphiniifolia and C. hastata in northern Japan. EDBD has cytotoxic effects and induces apoptosis via phosphorylation of p38 mitogen-activated protein kinase in human promyelocytic leukemia HL60 cells. However, the mechanism of action of EDBD has not yet been fully elucidated. In this study, we examined the molecular mechanisms of EDBD in the budding yeast Saccharomyces cerevisiae. EDBD arrested the growth of S. cerevisiae cells by suppressing progression from the G1 phase to the S phase and from the G2 phase to the M phase. Moreover, biochemical and genetic analyses revealed that EDBD activated environmental stress-response pathways involving Hog1 and affected Cln3/G1 cyclin activity, thereby inhibiting the expression of SCB-binding factor and MCB-binding factor target genes. Our results provided important insights into the functions of EDBD in tumor cells and may facilitate the development of EDBD-based antitumor therapies. •Swi4 physically interacts with Swi6 by anti tag coimmunoprecipitation (View interaction).
Sigma-Aldrich
Hydroxyurea, 98%, powder
Tiabendazole, European Pharmacopoeia (EP) Reference Standard
Hydroxycarbamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Thiabendazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Thiabendazole, PESTANAL®, analytical standard
Sigma-Aldrich
Thiabendazole, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Thiabendazole, ≥99%, powder
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture