Merck
  • Isomer-specific effects of conjugated linoleic acid on HDL functionality associated with reverse cholesterol transport.
Isomer-specific effects of conjugated linoleic acid on HDL functionality associated with reverse cholesterol transport.
The Journal of nutritional biochemistry (2014-12-04)
Nathalie Nicod, Robert S Parker, Elena Giordano, Virginia Maestro, Alberto Davalos, Francesco Visioli
ABSTRACT
High-density lipoproteins (HDLs) are atheroprotective because of their role in reverse cholesterol transport. The intestine is involved in this process because it synthesizes HDL, removes cholesterol from plasma and excretes it into the lumen. We investigated the role of selected dietary fatty acids on intestinal cholesterol uptake and HDL functionality. Caco-2 monolayers grown on Transwells were supplemented with either palmitic, palmitoleic, oleic, linoleic, docosahexaenoic, eicosapentaenoic, arachidonic or conjugated linoleic acids (CLAs): c9,t11-CLA; t9,t11-CLA; c10,t12-CLA. Cells synthesized HDL in the basolateral compartment for 24 h in the absence or presence of an antibody to SR-BI (aSR-BI), which inhibits its interaction with HDL. Free cholesterol (FC) accumulated to a greater extent in the presence than in the absence of aSR-BI, indicating net uptake of FC by SR-BI. Uptake's efficiency was significantly decreased when cells were treated with c9,t11-CLA relative to the other fatty acids. These differences were associated with lower HDL functionality, since neither SR-BI protein expression nor expression and alternative splicing of other genes involved lipid metabolism were affected. Only INSIG2 expression was decreased, with no increase of its target genes. Increasing pre-β-HDL synthesis, by inducing ABCA1 and adding APOA1, resulted in reduced uptake of FC by SR-BI after c9,t11-CLA treatment, indicating reduced functionality of pre-β-HDL. Conversely, treatment with c9,t11-CLA resulted in a greater uptake of FC and esterified cholesterol from mature HDL. Therefore, Caco-2 monolayers administered c9,t11-CLA produced a nonfunctional pre-β-HDL but took up cholesterol more efficiently via SR-BI from mature HDL.
Sigma-Aldrich
(L)-Dehydroascorbic acid
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Sodium taurocholate hydrate, ≥97.0% (TLC)
Sigma-Aldrich
Ergosterol, ≥75%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Dehydro-L-(+)-ascorbic acid dimer, ≥80% (enzymatic)
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ergosterol, Pharmaceutical Secondary Standard; Certified Reference Material
Ergosterol, European Pharmacopoeia (EP) Reference Standard
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurocholic acid sodium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
Taurocholic acid sodium salt hydrate, BioXtra, ≥95% (HPLC)