Merck
  • Agar films containing silver nanoparticles as new supports for electromembrane extraction.
Agar films containing silver nanoparticles as new supports for electromembrane extraction.
Analytical and bioanalytical chemistry (2014-12-19)
Cristina Román Hidalgo, María Ramos-Payán, Juan Antonio Ocaña-González, María Jesús Martín-Valero, Miguel Ángel Bello-López
ABSTRACT
A new support containing silver nanoparticles to assist electromembrane extraction (EME) procedures is proposed. For the first time, synthesized agar films containing silver nanoparticles (AgNPs) have been used as a support for liquid membranes in EME. Agarose films of 20 μm thickness containing 107.9 mg Ag/g agar were synthesized and characterized by transmission electron microscopy (TEM) and atomic force microscopy (AFM), showing isolated spherical silver nanoparticles of 20-30 nm diameter with homogeneous distribution. Nanometallic films were cut and glued to narrow bore glass tubes and used as supports for a dihexyl ether liquid membrane for use in an EME procedure. EME conditions were optimized and applied to the extraction of selected non-steroidal anti-inflammatory drugs (NSAIDs). The results were compared to those using polypropylene membranes (450 μm and 100 μm thickness), achieving 10- to 70-fold higher extraction efficiency. This article opens a new line of research into electrically assisted microextraction systems by combining other possible kinds of nanometallic films, including different metals, film functionalization through metallic NPs, and the use of low polarity solvents. Also, very low currents are obtained during the extraction process, which lead to high electromigration of the analytes.
Product Description
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(+)-L-Alliin, analytical standard
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-L-Alliin, ≥90% (HPLC)
Sigma-Aldrich
Naproxen, meets USP testing specifications
Naproxen, European Pharmacopoeia (EP) Reference Standard
Ketoprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ketoprofen, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ketoprofen, United States Pharmacopeia (USP) Reference Standard
USP
Naproxen, United States Pharmacopeia (USP) Reference Standard
Supelco
Ketoprofen, VETRANAL®, analytical standard
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Supelco
Naproxen, VETRANAL®, analytical standard
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Sigma-Aldrich
Dihexyl ether, 97%