Merck
  • L-carnitine attenuates H2O2-induced neuron apoptosis via inhibition of endoplasmic reticulum stress.
L-carnitine attenuates H2O2-induced neuron apoptosis via inhibition of endoplasmic reticulum stress.
Neurochemistry international (2014-09-16)
Junli Ye, Yantao Han, Xuehong Chen, Jing Xie, Xiaojin Liu, Shunhong Qiao, Chunbo Wang
ABSTRACT
Both oxidative stress and endoplasmic reticulum stress (ER stress) have been linked to pathogenesis of neurodegenerative diseases. Our previous study has shown that L-carnitine may function as an antioxidant to inhibit H2O2-induced oxidative stress in neuroblastoma SH-SY5Y cells. To further explore the neuroprotection of L-carnitine, here we study the effects of L-carnitine on the ER stress response in H2O2-induced SH-SY5Y cell injury. Our results showed that L-carnitine pretreatment could increase cell viability; inhibit apoptosis and ROS accumulation caused by H2O2 or tunicamycin (TM). L-carnitine suppress the endoplasmic reticulum dilation and activation of ER stress-associated proteins including glucose-regulated protein 78 (GRP78), CCAAT/enhancer-binding protein-homologous protein (CHOP), JNK, Bax and Bim induced by H2O2 or TM. In addition, H2O2-induced cell apoptosis and activation of ER stress can also be attenuated by antioxidant N-acetylcysteine (NAC), CHOP siRNA and the inhibitor of ER stress 4-phenylbutyric acid (4-PBA). Taken together, our results demonstrated that H2O2 could trigger both oxidative stress and ER stress in SH-SY5Y cells, and ER stress participated in SH-SY5Y apoptosis mediated by H2O2-induced oxidative stress. CHOP/Bim or JNK/Bim-dependent ER stress signaling pathways maybe related to the neuroprotective effects of L-carnitine against H2O2-induced apoptosis and oxidative injury.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
L-Carnitine inner salt, synthetic, ≥98%
Levocarnitine, European Pharmacopoeia (EP) Reference Standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
USP
Levocarnitine, United States Pharmacopeia (USP) Reference Standard
SAFC
Sodium deoxycholate