Merck
  • Irisin stimulates muscle growth-related genes and regulates adipocyte differentiation and metabolism in humans.
Irisin stimulates muscle growth-related genes and regulates adipocyte differentiation and metabolism in humans.
International journal of obesity (2005) (2014-03-13)
J Y Huh, F Dincer, E Mesfum, C S Mantzoros
ABSTRACT
Irisin is a recently identified exercise-induced myokine suggested to induce browning of white adipocytes. Deficiency of myostatin, and thus stimulation of muscle growth, has also been reported to induce irisin and its precursor FNDC5 expression in muscle and drive the browning of white adipocytes in mice, implying that irisin may be related to muscle growth in addition to its beneficial effects in adipocytes. In humans, the effect of irisin in muscle hypertrophy as well as adipocyte metabolism has not been fully investigated. Primary cultured human myocytes/adipocytes and 3T3-L1 cells were used to examine irisin-regulated gene/protein expression. Lipid accumulation, ATP content, glycolysis, lipolysis and metabolite profile were measured in control and irisin-treated (10 and 50 nM) adipocytes. In human myocytes, FNDC5 mRNA and irisin secretion were increased during myogenic differentiation, along with PGC1α and myogenin expression. Irisin treatment significantly increased insulin-like growth factor 1 and decreased myostatin gene expression through ERK pathway. PGC1α4, a newly discovered PGC1α isoform specifically related to muscle hypertrophy, was also upregulated. In human adipocytes, irisin induced uncoupling protein 1 and consequently increased adipocyte energy expenditure, expression of metabolic enzymes and metabolite intermediates, resulting in inhibition of lipid accumulation. Irisin and FNDC5 treatment also reduced preadipocyte differentiation, suggesting an additional mechanism in suppressing fat mass. These results suggest that irisin/FNDC5 has a pleiotropic role in muscle and improvement of adipocyte metabolism in humans.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Supelco
Glycerol, analytical standard