Merck
  • Association of SOD2 polymorphisms with primary open angle glaucoma in a Chinese population.
Association of SOD2 polymorphisms with primary open angle glaucoma in a Chinese population.
Ophthalmic genetics (2014-11-25)
Yu Zhou, Ping Shuai, Xiulan Li, Xiaoqi Liu, Jin Wang, Yin Yang, Fang Hao, He Lin, Dingding Zhang, Bo Gong
ABSTRACT
Genetic factors have been studied extensively for their role in the pathogenesis of primary open angle glaucoma (POAG). This study was conducted to investigate whether manganese superoxide dismutase (SOD2) variants play a significant role in POAG in a Chinese population. This study included of 416 unrelated POAG patients and 997 unrelated control subjects. Four SOD2 tag single nucleotide polymorphisms (SNPs), including rs6917589 rs2842980, rs5746136 and rs4880, were genotyped by dye terminator-based SNaPshot method. The genotype and allele frequencies were evaluated using the χ(2) tests. Allelic association analysis showed that there were suggestive differences in the allelic distributions between POAG cases and controls for SNPs rs6917589 and rs5746136 (p = 0.0.046 and p = 0.032, respectively), but no statistically significant association was detected between the two SNPs and POAG after Bonferroni correction (p > 0.0125). The allele and genotype frequency in SNPs rs2842980 and rs4880 showed no statistically significant difference between POAG cases and controls (p = 0.128 and p = 0.867, respectively). SNP rs5746136 had a significant association with POAG in the recessive model (p = 0.003155). Haplotype ATGT generated from the four SNPs showed a trend of association with POAG (p = 0.0098). Our results showed a trend of association with POAG, suggesting that SOD2 may play a significant role in the development of POAG in the Chinese population. Further work with a larger sample size and functional study is needed to confirm the importance of the SOD2 gene in the pathogenesis of glaucoma.
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol