Merck
  • An evaluation of acute hydrogen sulfide poisoning in rats through serum metabolomics based on gas chromatography-mass spectrometry.
An evaluation of acute hydrogen sulfide poisoning in rats through serum metabolomics based on gas chromatography-mass spectrometry.
Chemical & pharmaceutical bulletin (2014-06-03)
Meiling Zhang, Mingjie Deng, Jianshe Ma, Xianqin Wang
ABSTRACT
Hydrogen sulfide (H2S) is the second leading cause of toxin-related deaths in the operational site. Its main target organs of toxic effects are the central nervous system and respiratory system. In this study, we developed a serum metabonomic method, based on gas chromatography-mass spectrometry (GC/MS), to evaluate the effect of acute poisoning by hydrogen sulfide on rats. Pattern recognition analysis, including both principal component analysis (PCA) and partial least squares-discriminate analysis (PLS-DA), revealed that acute hydrogen sulfide poisoning induced metabolic perturbations. Compared to the control group, the level of urea, glucose, glyceryl stearate in rat serum of the poisoning group increased after two hours, and the level of glucose, docosahexaenoic acid, glyceryl stearate and arachidonic acid in rat serum of the poisoning group increased after 48 h, while the L-valine, galactose, L-tyrosine levels decreased. Our results indicate that metabonomic methods based on GC/MS may be useful to elucidate acute hydrogen sulfide poisoning through the exploration of biomarkers.
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
N-Methylhydroxylamine hydrochloride, 98%
Sigma-Aldrich
Trifluoroacetamide, 97%
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ≥99%
Supelco
Pyridine, analytical standard
Supelco
Acetonitrile, analytical standard
Supelco
Acetic anhydride, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Supelco
N-Methyl-bis(trifluoroacetamide), derivatization grade (GC derivatization), LiChropur, ≥97.0% (GC)
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Heptane, anhydrous, 99%