Merck
  • Dental pulp stem cells immobilized in alginate microspheres for applications in bone tissue engineering.
Dental pulp stem cells immobilized in alginate microspheres for applications in bone tissue engineering.
International endodontic journal (2013-10-17)
M M Kanafi, A Ramesh, P K Gupta, R R Bhonde
ABSTRACT
To immobilize dental pulp stem cells (DPSC) in alginate microspheres and to determine cell viability, proliferation, stem cell characteristics and osteogenic potential of the immobilized DPSCs. Human DPSCs isolated from the dental pulp were immobilized in 1% w/v alginate microspheres. Viability and proliferation of immobilized DPSCs were determined by trypan blue and MTT assay, respectively. Stem cell characteristics of DPSCs post immobilization were verified by labelling the cells with CD73 and CD90. Osteogenic potential of immobilized DPSCs was assessed by the presence of osteocalcin. Alizarin red staining and O-cresolphthalein complexone method confirmed and quantified calcium deposition. A final reverse transcriptase PCR evaluated the expression of osteogenic markers - ALP, Runx-2 and OCN. More than 80% of immobilized DPSCs were viable throughout the 3-week study. Proliferation appeared controlled and consistent unlike DPSCs in the control group. Presence of CD73 and CD90 markers confirmed the stem cell nature of immobilized DPSCs. The presence of osteocalcin, an osteoblastic marker, was confirmed in the microspheres on day 21. Mineralization assays showed high calcium deposition indicating elevated osteogenic potential of immobilized DPSCs. Osteogenic genes- ALP, Runx-2 and OCN were also upregulated in immobilized DPSCs. Surprisingly, immobilized DPSCs in the control group cultured in conventional stem cell media showed upregulation of osteogenic genes and expressed osteocalcin. Dental pulp stem cells immobilized in alginate hydrogels exhibit enhanced osteogenic potential while maintaining high cell viability both of which are fundamental for bone tissue regeneration.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Sigma-Aldrich
o-Cresolphthalein Complexone, powder
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)