Merck
  • Affinity precipitation of a monoclonal antibody from an industrial harvest feedstock using an ELP-Z stimuli responsive biopolymer.
Affinity precipitation of a monoclonal antibody from an industrial harvest feedstock using an ELP-Z stimuli responsive biopolymer.
Biotechnology and bioengineering (2014-03-07)
Rahul D Sheth, Mi Jin, Bharat V Bhut, Zhengjian Li, Wilfred Chen, Steven M Cramer
ABSTRACT
In this work, a proof of concept elastin-like polypeptide-Z domain fusion (ELP-Z) based affinity precipitation process is developed for monoclonal antibody (mAb) purification from industrial harvest feeds. Greater than 99% mAb recoveries are obtained during the initial binding step of the process for both pure mAb and the mAb harvest feeds. Great than 90% overall mAb yields are also obtained for the subsequent elution step of the process with no measurable mAb aggregation. The process is shown to result in more than 2 logs of host cell protein (HCP) and more than 4 logs of DNA clearance from the harvest feed. While the overall mAb yield and HCP clearance for the affinity precipitation process was comparable to Protein A chromatography the DNA clearance was clearly superior. Performance is maintained for mAb final elution concentrations up to 20 g/L, demonstrating the ability of the process to both concentrate and purify the mAb. Effective ELP-Z regeneration is also demonstrated using 0.1 M NaOH with no adverse effect on subsequent capture efficiency. Finally, the reusability of the ELP-Z construct and robustness of the process is demonstrated for up to three purification-regeneration cycles with minimal product and impurity carryover and high yields and purity. This work demonstrates that the ELP-Z based precipitation approach can be successfully employed as an affinity capture step for industrial mAbs.
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
L-Arginine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O