Merck
  • Efficacy and safety of triple combination therapy with artesunate-amodiaquine-methylene blue for falciparum malaria in children: a randomized controlled trial in Burkina Faso.
Efficacy and safety of triple combination therapy with artesunate-amodiaquine-methylene blue for falciparum malaria in children: a randomized controlled trial in Burkina Faso.
The Journal of infectious diseases (2014-10-01)
Boubacar Coulibaly, Michael Pritsch, Mamadou Bountogo, Peter E Meissner, Eric Nebié, Christina Klose, Meinhard Kieser, Nicole Berens-Riha, Andreas Wieser, Sodiomon B Sirima, Jörg Breitkreutz, R Heiner Schirmer, Ali Sié, Frank P Mockenhaupt, Chris Drakeley, Teun Bousema, Olaf Müller
ABSTRACT
Methylene blue (MB) has been shown to be safe and effective against falciparum malaria in Africa and to have pronounced gametocytocidal properties. Three days of treatment with artesunate (AS)-amodiaquine (AQ) combined with MB was compared with AS-AQ treatment in a randomized controlled phase IIb study; the study included 221 children aged 6-59 months with uncomplicated falciparum malaria in Burkina Faso. The primary end point was gametocyte prevalence during follow-up, as determined by microscopy and real-time quantitative nucleic acid sequence-based amplification (QT-NASBA). The gametocyte prevalence of Plasmodium falciparum at baseline was 3.6% (microscopy) and 97% (QT-NASBA). It was significantly lower in the AS-AQ-MB than in the AS-AQ group on day 7 of follow-up (microscopy, 1.2% vs 8.9% [P < .05]; QT-NASBA, 36.7% vs 63.3% [P < .001]). Hemoglobin values were significantly lower in the AS-AQ-MB group than in the AS-AQ group at days 2 and 7 of follow-up. Vomiting of the study medication occurred significantly more frequently in the AS-AQ-MB group. The combination of MB with an artemisinin-based combination therapy has been confirmed to be effective against the gametocytes of P. falciparum. MB-based combinations need to be compared with primaquine-based combinations, preferably using MB in an improved pediatric formulation. Clinical Trials Registration: NCT01407887.
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucralose, United States Pharmacopeia (USP) Reference Standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sucralose, European Pharmacopoeia (EP) Reference Standard
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucralose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Sucralose, analytical standard