Merck
  • Development and validation of method for TH588 and TH287, potent MTH1 inhibitors and new anti-cancer agents, for pharmacokinetic studies in mice plasma.
Development and validation of method for TH588 and TH287, potent MTH1 inhibitors and new anti-cancer agents, for pharmacokinetic studies in mice plasma.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Aljona Saleh, Camilla Gökturk, Ulrika Warpman-Berglund, Thomas Helleday, Ingrid Granelli
ABSTRACT
MTH1 is a protein that is required for cancer cell survival and is overexpressed in cancer cells. TH588 and TH287 are two new compounds that inhibit the MTH1 protein. The inhibitors were tested in pharmacokinetic studies on mice. A bioanalytical method was developed and validated for determination in mice plasma. The method was based on protein precipitation followed by LC-MS/MS analysis. The separation was performed on an Ascentis Express RP-Amide C18 column. The mass spectrometer was operated in positive electrospray ionization mode and the analytes were determined with multiple reaction monitoring (MRM). Abundant monoisotopic fragments were used for quantification. Two additional fragments were used for conformational analysis. The recovery of the compounds in plasma varied between 61 and 91% and the matrix effects were low and ranged between -3% and +2%. The method showed to be selective, linear, accurate and precise, and applicable for preclinical pharmacokinetic studies of TH588 and TH287 in mouse plasma. Half-life (T1/2) was ≤3.5h and maximum concentration (Cmax) ranged between 0.82 and 338μM for the different administration routes and compounds.
Sigma-Aldrich
Ultrapure Acetonitrile
Supelco
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Ascentis® Express RP-Amide, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® Express RP-Amide, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 3 mm
Supelco
Ascentis® Express RP-Amide, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express RP-Amide, 2 μm Guard Cartridge, 2 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
Supelco
Ascentis® Express RP-Amide, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® Express RP-Amide, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
Ascentis® Express RP-Amide, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 2.1 mm
Supelco
Ascentis® Express RP-Amide, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 2.1 mm
Supelco
Ascentis® Express RP-Amide, 2 μm Guard Cartridge, 2 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
Supelco
Ascentis® Express RP-Amide, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 2.1 mm
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Supelco
Ascentis® Express RP-Amide, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Ascentis® Express RP-Amide, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 3 mm
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 2 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 10 mm, pk of 1 ea
Supelco
Ascentis® Express RP-Amide, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 4.6 mm, pk of 1 ea