Merck
  • Galactosylated bovine serum albumin nanoparticles for parenteral delivery of oridonin: tissue distribution and pharmacokinetic studies.
Galactosylated bovine serum albumin nanoparticles for parenteral delivery of oridonin: tissue distribution and pharmacokinetic studies.
Journal of microencapsulation (2014-04-05)
Caiyun Li, Dianrui Zhang, Yuanyuan Guo, Hejian Guo, Tingting Li, Leilei Hao, Dandan Zheng, Guangpu Liu, Qiang Zhang
ABSTRACT
Bovine serum albumin (BSA) nanoparticle is a promising drug carrier system. Oridonin (ORI)-loaded galactosylated BSA nanoparticle (ORI-GB-NP) was prepared for liver targeting delivery of ORI. This work was designed to investigate the in vitro release, in vivo pharmacokinetics and tissue distribution of ORI-GB-NP. ORI-GB-NP was prepared by the desolvation method. The particle size of ORI-GB-NP was 172.0 ± 8.3 nm with narrow size distribution. The in vitro release of ORI-GB-NP exhibited biphasic drug release pattern with an initial burst release and consequently sustained release. Pharmacokinetic analysis displayed that ORI-GB-NP and ORI-loaded BSA nanoparticle (ORI-BSA-NP) could enhance the drug plasma level and prolong the circulation time in contrast with ORI solution. Meanwhile, compared with ORI-BSA-NP, ORI-GB-NP could deliver more ORI to liver and simultaneously reduce the toxicity of ORI to heart, lung and kidney. In conclusion, ORI-GB-NP could be a promising drug delivery system for liver cancer therapy.
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC