Merck
  • An airway epithelial iNOS-DUOX2-thyroid peroxidase metabolome drives Th1/Th2 nitrative stress in human severe asthma.
An airway epithelial iNOS-DUOX2-thyroid peroxidase metabolome drives Th1/Th2 nitrative stress in human severe asthma.
Mucosal immunology (2014-02-13)
N Voraphani, M T Gladwin, A U Contreras, N Kaminski, J R Tedrow, J Milosevic, E R Bleecker, D A Meyers, A Ray, P Ray, S C Erzurum, W W Busse, J Zhao, J B Trudeau, S E Wenzel
ABSTRACT
Severe refractory asthma is associated with enhanced nitrative stress. To determine the mechanisms for high nitrative stress in human severe asthma (SA), 3-nitrotyrosine (3NT) was compared with Th1 and Th2 cytokine expression. In SA, high 3NT levels were associated with high interferon (IFN)-γ and low interleukin (IL)-13 expression, both of which have been reported to increase inducible nitric oxide synthase (iNOS) in human airway epithelial cells (HAECs). We found that IL-13 and IFN-γ synergistically enhanced iNOS, nitrite, and 3NT, corresponding with increased H(2)O(2). Catalase inhibited whereas superoxide dismutase enhanced 3NT formation, supporting a critical role for H(2)O(2), but not peroxynitrite, in 3NT generation. Dual oxidase-2 (DUOX2), central to H(2)O(2) formation, was also synergistically induced by IL-13 and IFN-γ. The catalysis of nitrite and H(2)O(2) to nitrogen dioxide radical (NO(2)(•)) requires an endogenous peroxidase in this epithelial cell system. Thyroid peroxidase (TPO) was identified by microarray analysis ex vivo as a gene distinguishing HAEC of SA from controls. IFN-γ induced TPO in HAEC and small interfering RNA knockdown decreased nitrated tyrosine residues. Ex vivo, DUOX2, TPO, and iNOS were higher in SA and correlated with 3NT. Thus, a novel iNOS-DUOX2-TPO-NO(2)(•) metabolome drives nitrative stress in HAEC and likely in SA.
Product Description
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
MISSION® esiRNA, targeting human TPO
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized