Merck
  • Statins combined with exercise are associated with the increased renal function mediated by high-molecular-weight adiponectin in coronary artery disease patients.
Statins combined with exercise are associated with the increased renal function mediated by high-molecular-weight adiponectin in coronary artery disease patients.
Journal of cardiology (2014-01-25)
Kensuke Toyama, Seigo Sugiyama, Hideki Oka, Yuri Iwasaki, Hitoshi Sumida, Tomoko Tanaka, Shinji Tayama, Hideaki Jinnouchi, Hisao Ogawa
ABSTRACT
Statins and exercise therapy are clinically effective in preventing cardiovascular events in patients with coronary artery disease (CAD). The aim of this study was to determine the effects of statins combined with exercise on the renal function of CAD patients. We performed a sub-analysis of a clinical trial that determined the 20-week-effects of two statins (rosuvastatin, n=14; atorvastatin, n=14) combined with regular exercise on renal function, as assessed by the estimated glomerular filtration rates (eGFRs) of CAD patients. The combination of statins and exercise therapy increased eGFRs from 61.1±16.6 at baseline to 65.8±16.8ml/min/per 1.73m(2) (p=0.03), increased serum levels of high-molecular-weight (HMW) adiponectin, increased ubiquinol/low-density lipoprotein cholesterol (LDL-C) ratios, and decreased high sensitivity C-reactive protein (hs-CRP). Changes in HMW-adiponectin, ubiquinol/LDL-C ratios and hs-CRP were significantly correlated with changes in eGFR (r=0.597, p=0.001; r=0.437, p=0.02; and r=-0.473, p=0.01, respectively). Treatment-induced increases in HMW-adiponectin independently correlated with the increases in eGFR (β=0.513, p=0.02) in a multivariate analysis. Both atorvastatin and rosuvastatin combined with regular exercise produced increases in eGFR. The patients treated with rosuvastatin exhibited significant improvements in eGFR. Statins combined with exercise significantly increased eGFR in CAD patients, and these improvements in renal function were correlated with increases in HMW-adiponectin levels. The statins-exercise combination treatment may have provided clinical benefits for patients with CAD partly through the improvement in renal function.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications