Merck
  • Metabolic discrimination of Swertia mussotii and Swertia chirayita known as "Zangyinchen" in traditional Tibetan medicine by (1)H NMR-based metabolomics.
Metabolic discrimination of Swertia mussotii and Swertia chirayita known as "Zangyinchen" in traditional Tibetan medicine by (1)H NMR-based metabolomics.
Journal of pharmaceutical and biomedical analysis (2014-07-06)
Gang Fan, Wei-Zao Luo, Shang-Hua Luo, Yan Li, Xian-Li Meng, Xiang-Dong Zhou, Yi Zhang
ABSTRACT
Swertia mussotii Franch. and Swertia chirayita Buch.-Ham. have been commonly used under the same name "Zangyinchen" for the treatment of liver and gallbladder diseases in traditional Tibetan medicine. Detailed characterization and comparison of the complete set of metabolites of these two species are critical for their objective identification and quality control. In this study, a rapid, simple and comprehensive (1)H NMR-based metabolomics method was first developed to differentiate the two species. A broad range of metabolites, including iridoid glycosides, xanthones, triterpenoids, flavonoids, carbohydrates, and amino acids, were identified. Statistical analysis showed evident differences between the two species, and the major markers responsible for the differences were screened. In addition, quantitative (1)H NMR method (qHNMR) was used for the target analysis of the discriminating metabolites. The results showed that S. mussotii had significantly higher contents of gentiopicrin, isoorientin, glucose, loganic acid, and choline, whereas S. chirayita exhibited higher levels of swertiamarin, oleanolic acid, valine, and fatty acids. These findings indicate that (1)H NMR-based metabolomics is a reliable and effective method for the metabolic profiling and discrimination of the two Swertia species, and can be used to verify the genuine origin of Zangyinchen.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, anhydrous, ≥99.8 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, "100%", 99.96 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, 99 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D