Merck
  • In vitro antifungal activity and probable fungicidal mechanism of aqueous extract of Barleria grandiflora.
In vitro antifungal activity and probable fungicidal mechanism of aqueous extract of Barleria grandiflora.
Applied biochemistry and biotechnology (2015-02-13)
Suman Kumari, Preeti Jain, Bhawana Sharma, Preeti Kadyan, Rajesh Dabur
ABSTRACT
Barleria grandiflora Dalz. (Acanthaceae) is being used in India to treat different types of disorders including skin infections. Therefore, there are good possibilities to find antifungal compounds in its extracts with novel mechanism of action. The main objectives of the present study were to evaluate the antifungal activity of plant extracts and to study its effects on metabolic pathways of A. fumigatus. The microbroth dilution assay was used to explore antifungal activity and MIC of various extracts. Metabolic profiles of control and treated cultures were collected from Q-TOF-MS interfaced with HPLC. Affected metabolic pathways of A. fumigatus after the treatment were analyzed by discrimination analysis of mass data. Antifungal activities were observed in hot and cold water extracts of the plant. Hot water extract of B. grandiflora showed significant activity against tested fungi in the range 0.625-1.25 mg/mL. Partial least discrimination analysis revealed that the hot water plant extract downregulated amino acid, glyoxylate pathway, and methylcitrate pathways at the same time due to the synergistic effects of secondary metabolites. Hot water extract also downregulated several other metabolic pathways unique to fungi indicating its specific activity toward fungi. B. grandiflora showed promising antifungal activity which can further be exploited by identification of active compounds, to inhibit the specific fungal pathways and development of novel therapeutic antifungal drugs.
Product Description
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Camphor (dl), primary reference standard
Camphor (racemic), European Pharmacopoeia (EP) Reference Standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ultrapure Acetonitrile
Supelco
D-Camphor, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
D-Camphor, ≥97%, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
D-Camphor, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard