Merck
  • B-cell activating factor-receptor specific activation of tumor necrosis factor receptor associated factor 6 and the phosphatidyl inositol 3-kinase pathway in lymphoma B cells.
B-cell activating factor-receptor specific activation of tumor necrosis factor receptor associated factor 6 and the phosphatidyl inositol 3-kinase pathway in lymphoma B cells.
Leukemia & lymphoma (2013-11-12)
Frank Secreto, Michelle Manske, Tammy Price-Troska, Steven Ziesmer, Lucy S Hodge, Stephen M Ansell, James R Cerhan, Anne J Novak
ABSTRACT
B-cell activating factor-receptor (BAFF-R) is the primary BAFF receptor that is responsible for promoting B-cell development and survival. Malignant B-cells exploit the BAFF/BAFF-R system, and high serum BAFF levels or genetic alterations in BAFF receptors have been found in B-cell cancers. BAFF signaling impacts pro-survival pathways. However, other than nuclear factor-κB2 (NF-κB2), little is known about the specific pathways activated by individual BAFF receptors. Using a novel BAFF-R expression model we have demonstrated that activation of BAFF-R, independent of transmembrane activator and cytophilin ligand interactor (TACI) and B-cell maturation antigen (BCMA), can induce phosphorylation of Akt and glycogen synthase kinase 3β (GSK3β). Expression of an activated form of BAFF-R also enhanced a pro-survival gene expression pattern, including the novel BAFF-regulated gene Pin1, whose expression was phosphatidyl inositol 3-kinase (PI3K)-dependent. Additionally, we showed that TRAF6 is essential for mediating BAFF-R dependent activation of Akt. Together these data describe a novel role for TRAF6 in BAFF-R-specific activation of the PI3K pathway and provide evidence suggesting a new role for Pin1 in BAFF-R signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Traf6
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
MISSION® esiRNA, targeting human TRAF6
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)