Merck
  • Amlodipine protects rat ventricular cardiomyoblast H9c2 From hypoxia-induced apoptosis and restores oxidative balance by Akt-1-dependent manner.
Amlodipine protects rat ventricular cardiomyoblast H9c2 From hypoxia-induced apoptosis and restores oxidative balance by Akt-1-dependent manner.
Journal of cardiovascular pharmacology (2014-06-13)
Sarita Nehra, Varun Bhardwaj, Deepika Saraswat
ABSTRACT
Hypoxia-induced rise in intracellular calcium concentration is a causative agent of apoptosis and oxidative damage in cardiomyocytes. We examined the efficacy of calcium channel blocker amlodipine in preventing hypoxia-induced apoptosis in H9c2 cells and restoring oxidative balance. H9c2 cells were exposed to hypoxia (0.5% oxygen) to evaluate the efficacy of amlodipine in restoring cellular calcium levels. Cellular markers of apoptosis (Bax/Bcl2 and caspase-3, -7, and -9) and pro-survival markers (Akt/p-Akt levels) were evaluated under hypoxia. Redox damage was evaluated by assessing markers of oxidative damage, that is, glutathione reduced, glutathione oxidized, lipid peroxidation, reactive oxygen species, and manganese superoxide dismutase activity. Cellular adenosine triphosphate (ATP) pool and AMPKα levels were measured to evaluate regulation of metabolism under hypoxia. Amlodipine treatment at 25 nM prevented apoptosis and restored cellular calcium levels and oxidative damage in cardiomyocytes. Stabilization of caspase-3, -7, and -9 along with restoration of Akt/p-Akt levels depicted pro-survival efficacy of amlodipine. Also, restoration of cellular ATP and AMPKα levels indicates that amlodipine prevents cardiomyocytes from hypoxia-induced metabolic stress. Amlodipine thus protects H9c2 cells from hypoxia-induced apoptosis by regulating Akt/p-Akt-mediated caspase-3, -7, and -9 activation and restoring cellular ATP and redox status.
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
L-Glutathione oxidized disodium salt, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder