Merck
  • Analysis of telomerase activity based on a spired DNA tetrahedron TS primer.
Analysis of telomerase activity based on a spired DNA tetrahedron TS primer.
Biosensors & bioelectronics (2014-09-10)
Yan Li, Yanli Wen, Lele Wang, Wen Liang, Li Xu, Shuzhen Ren, Ziying Zou, Xiaolei Zuo, Chunhai Fan, Qing Huang, Gang Liu, Nengqin Jia
ABSTRACT
The development of sensitive telomerase biosensors is hindered by the restricted accessibility of telomere strand (TS) primer and the limited enzyme reaction space, which is mainly confined by the vertical distance. In this work, we designed an electrochemical telomerase biosensor based on a spired DNA tetrahedron TS primer (STTS). By adding a rigid dsDNA spire onto the top of the DNA tetrahedron, we successfully regulated the distance between the TS primer and the surface, and thus greatly facilitated the telomerase elongation on surface. The signal-to-noise ratio was 2 times higher than TSP without the spire structure. The limit of detection was calculated to be lower than 10 HeLa cells, which is at least 2 magnitudes lower than other surface extension-based electrochemical telomerase sensors without amplification. The practicability of STTS sensor was also demonstrated by analysing various other cell lines including cancer cells, stem cells of high telomerase activity and somatic cells of low telomerase activity.
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
7-Amino-4-hydroxy-2-naphthalenesulfonic acid, technical, ≥90% (T)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)