Merck
  • Anti-high mobility group box 1 (anti-HMGB1) antibodies are not related to the occurrence of cutaneous lesions in systemic lupus erythematosus.
Anti-high mobility group box 1 (anti-HMGB1) antibodies are not related to the occurrence of cutaneous lesions in systemic lupus erythematosus.
Scandinavian journal of rheumatology (2014-12-03)
G Tan, N Zhu, Z Shi, Z Meng, M Yu, K Li, J Yin, K Wei, X Mi, L Wang
ABSTRACT
To investigate whether serum anti-high mobility group box 1 (anti-HMGB1) antibodies are related to the development of skin lesions in systemic lupus erythematosus (SLE). This study involved 21 SLE patients with skin lesions, 18 without skin lesions, and 22 healthy controls. The presence and serum levels of anti-HMGB1-IgG and -IgM were measured by western blot and enzyme-linked immunosorbent assay (ELISA), respectively. HMGB1 expression and serum antibodies deposited in the skin were visualized by immunofluorescence staining. Using western blot analysis, we detected anti-HMGB1-IgG antibodies in 13 out of 21 SLE patients with skin lesions and 11 out of 18 SLE patients without skin lesions (p > 0.05). Serum levels of anti-HMGB1-IgG measured by ELISA were also comparable between the two groups of SLE patients (p > 0.05) but were higher in patients than in healthy controls (p < 0.05). Similar results were found with serum anti-HMGB1-IgM antibodies. HMGB1 accumulated under the stratum corneum in lupus cutaneous lesions without forming immune complexes with IgG or IgM, which were mainly observed along the epidermal-dermal junction. Furthermore, serum anti-HMGB1-IgM was higher in the group of patients with arthritis than in those without arthritis. Based on the findings of the current study, it is unlikely that anti-HMGB1 antibodies play a role in the development of SLE cutaneous lesions.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard