Merck
  • Nontargeted metabolite profiling discriminates diet-specific biomarkers for consumption of whole grains, fatty fish, and bilberries in a randomized controlled trial.
Nontargeted metabolite profiling discriminates diet-specific biomarkers for consumption of whole grains, fatty fish, and bilberries in a randomized controlled trial.
The Journal of nutrition (2014-12-21)
Kati Hanhineva, Maria A Lankinen, Anna Pedret, Ursula Schwab, Marjukka Kolehmainen, Jussi Paananen, Vanessa de Mello, Rosa Sola, Marko Lehtonen, Kaisa Poutanen, Matti Uusitupa, Hannu Mykkänen
ABSTRACT
Nontargeted metabolite profiling allows for concomitant examination of a wide range of metabolite species, elucidating the metabolic alterations caused by dietary interventions. The aim of the current study was to investigate the effects of dietary modifications on the basis of increasing consumption of whole grains, fatty fish, and bilberries on plasma metabolite profiles to identify applicable biomarkers for dietary intake and endogenous metabolism. Metabolite profiling analysis was performed on fasting plasma samples collected in a 12-wk parallel-group intervention with 106 participants with features of metabolic syndrome who were randomly assigned to 3 dietary interventions: 1) whole-grain products, fatty fish, and bilberries [healthy diet (HD)]; 2) a whole-grain-enriched diet with the same grain products as in the HD intervention but with no change in fish or berry consumption; and 3) refined-wheat breads and restrictions on fish and berries (control diet). In addition, correlation analyses were conducted with the food intake data to define the food items correlating with the biomarker candidates. Nontargeted metabolite profiling showed marked differences in fasting plasma after the intervention diets compared with the control diet. In both intervention groups, a significant increase was observed in 2 signals identified as glucuronidated alk(en)-ylresorcinols [corrected P value (Pcorr) < 0.05], which correlated strongly with the intake of whole-grain products (r = 0.63, P < 0.001). In addition, the HD intervention increased the signals for furan fatty acids [3-carboxy-4-methyl-5-propyl-2-furanpropionic acid (CMPF)], hippuric acid, and various lipid species incorporating polyunsaturated fatty acids (Pcorr < 0.05). In particular, plasma CMPF correlated strongly with the intake of fish (r = 0.47, P < 0.001) but not with intakes of any other foods. Novel biomarkers of the intake of health-beneficial food items included in the Nordic diet were identified by the metabolite profiling of fasting plasma and confirmed by the correlation analyses with dietary records. The one with the most potential was CMPF, which was shown to be a highly specific biomarker for fatty fish intake. This trial was registered at clinicaltrials.gov as NCT00573781.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ultrapure Acetonitrile
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%