Merck
  • microRNA-495 inhibits chondrogenic differentiation in human mesenchymal stem cells by targeting Sox9.
microRNA-495 inhibits chondrogenic differentiation in human mesenchymal stem cells by targeting Sox9.
Stem cells and development (2014-03-25)
Seulgi Lee, Dong Suk Yoon, Seungil Paik, Kyoung-Mi Lee, Yeonsue Jang, Jin Woo Lee
ABSTRACT
The chondrogenic differentiation process of human mesenchymal stem cells (hMSCs) passes through multiple stages, which are carried out by various factors and their interactions. Recently, microRNAs that regulate chondrogenic differentiation have been reported. However, microRNA that regulates SRY-related high mobility group-box gene 9 (Sox9), a chondrogenic key factor, has not been identified in hMSC. In this study, we identified that microRNA-495 (miR-495) is an important regulator of hMSC chondrogenic differentiation. In our microarray, miR-495 was downregulated during transforming growth factor (TGF)-β3-induced chondrogenic differentiation of hMSCs in vitro. We found that there is an miR-495 binding site in the 3' untranslated region (3'UTR) of Sox9. We confirmed opposite expression between miR-495 and Sox9 by using real-time polymerase chain reaction. Further, overexpression of miR-495 inhibited Sox9 expression, and repression of miR-495 increased expression of Sox9 in SW1353 cells and hMSCs. Additionally, luciferase analysis revealed that miR-495 directly binds to the Sox9 3'UTR, and we confirmed a seed sequence of miR-495 on the Sox9 3'UTR. Subsequently, overexpression of miR-495 repressed the expression of the extracellular matrix (ECM) protein, such as type II collagen (Col2A1), aggrecan, and proteoglycan products, whereas inhibition of miR-495 increased their expression. Collectively, this study indicates that miR-495 directly targets Sox9, ultimately leading to the regulation of chondrogenic differentiation in hMSCs.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Gram′s safranin solution, suitable for microscopy
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Safranin O, certified by the BSC
Sigma-Aldrich
Safranin O, Dye content ≥85 %
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Safranin O, suitable for microscopy