Merck
  • Quantitative comparison of a human cancer cell surface proteome between interphase and mitosis.
Quantitative comparison of a human cancer cell surface proteome between interphase and mitosis.
The EMBO journal (2014-12-06)
Nurhan Özlü, Mohammad H Qureshi, Yusuke Toyoda, Bernhard Y Renard, Gürkan Mollaoglu, Nazlı E Özkan, Selda Bulbul, Ina Poser, Wiebke Timm, Anthony A Hyman, Timothy J Mitchison, Judith A Steen
ABSTRACT
The cell surface is the cellular compartment responsible for communication with the environment. The interior of mammalian cells undergoes dramatic reorganization when cells enter mitosis. These changes are triggered by activation of the CDK1 kinase and have been studied extensively. In contrast, very little is known of the cell surface changes during cell division. We undertook a quantitative proteomic comparison of cell surface-exposed proteins in human cancer cells that were tightly synchronized in mitosis or interphase. Six hundred and twenty-eight surface and surface-associated proteins in HeLa cells were identified; of these, 27 were significantly enriched at the cell surface in mitosis and 37 in interphase. Using imaging techniques, we confirmed the mitosis-selective cell surface localization of protocadherin PCDH7, a member of a family with anti-adhesive roles in embryos. We show that PCDH7 is required for development of full mitotic rounding pressure at the onset of mitosis. Our analysis provided basic information on how cell cycle progression affects the cell surface. It also provides potential pharmacodynamic biomarkers for anti-mitotic cancer chemotherapy.
Product Description
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Diethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Supelco
Diethylene glycol, analytical standard
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)