Merck
  • Montelukast-loaded nanostructured lipid carriers: part II pulmonary drug delivery and in vitro-in vivo aerosol performance.
Montelukast-loaded nanostructured lipid carriers: part II pulmonary drug delivery and in vitro-in vivo aerosol performance.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-08-01)
Arpana Patil-Gadhe, Abhay Kyadarkunte, Milind Patole, Varsha Pokharkar
ABSTRACT
The aim of the present study was to establish the potential of montelukast loaded nanostructured lipid carrier (MNLC) for pulmonary application. The formulated nanoparticles were evaluated in vitro for aerodynamic characterization and in vivo for pulmokinetics in Wistar rats. The in vitro cytotoxicity was performed on A549 cell line and compared with montelukast-aqueous solution. MNLC was prepared with montelukast (0.2%), Precirol ATO5 (solid lipid), and Capryol-90 (liquid lipid) in the ratio of 7:3 using melt-emulsification-homogenization method. dl-Pyrrolidonecarboxylic acid salt of l-cocyl arginine ethyl ester (CAE), a biodegradable surfactant in the concentration of 1% was used to stabilize the nanoparticles. The particle size and encapsulation efficiency (EE) were 184.6 ± 2.7 nm and >95%, respectively. MNLC-Dry powder for inhalation (DPI) was prepared by lyophilization using 3% mannitol as cryoprotectant and carrier. MNLC-DPI was evaluated for flow, crystallographic and thermal properties. Mass median diameters (MMD) and density for MNLC-DPI were found to be 15.1 ± 1.4 μm and 0.051 ± 0.002 g/cc, respectively. In vitro aerosol performance study indicated more than 95% of the emitted dose (ED) at both the flow rates studied. Mass median aerodynamic diameters (MMAD) of 3.24 ± 0.67 μm with 69.98 ± 1.9% fine particle fraction (FPF) were obtained at 30 L/min flow rate, whereas at 60 L/min MMAD and FPF were found to be 2.83 ± 0.46 μm and 90.22 ± 2.6%, respectively. In vitro cytotoxicity study on A549 cells revealed higher safety of MNLC than pure drug. The pulmonary pharmacokinetic study demonstrated improved bioavailability, longer residence of drug in the lung and targeting factor of 11.76 for MNLC as compared to montelukast-aqueous solution. Thus, the results of the study demonstrated the potential of montelukast lipidic nanoparticulate formulation to improve the efficacy with reduced toxicity leading to better performance of drug as MNLC-DPI for inhalation use.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard