Merck
  • Anti-diabetic and spasmolytic potential of Farsetia hamiltonii Royle from Cholistan desert.
Anti-diabetic and spasmolytic potential of Farsetia hamiltonii Royle from Cholistan desert.
Journal of ethnopharmacology (2014-09-16)
Muhammad Munawar Hayat, Sadia Sarwar, Shazia Anjum, Muhammad Uzair, Hafiz Muhammad Farhan Rasheed, Qaiser Jabeen, Bashir Ahmad Choudhary, Muhammad Ashraf
ABSTRACT
Folk herbal practitioners of the Cholistan desert claim Farsetia hamiltonii Royle (Brassicaceae) to treat diabetes, oxidative damages, diarrhea, fever, and abdominal cramps. The aim of this study was to scientifically find the potential of Farsetia hamiltonii in treating diabetes and gastrointestinal diseases. In vivo anti-diabetic activity of Farsetia hamiltonii was studied on alloxan induced diabetic rats to justify its traditional use. The in vitro antispasmodic activity on isolated tissues of rabbit jejunum was also evaluated. In addition, several enzyme inhibition studies (lipoxygenase, tyrosinase, acetylcholinesterase (AchE), carbonic II anhydrase and phosphodiesterase I) and antioxidant activity of plant extracts were also conducted. In vivo experiments, Farsetia hamiltonii methanol extract (300 mg/kg) significantly lowered the fasting blood glucose (107.6 ± 1.249 mg/dL up to 4th day) comparable to positive control (Glibenclamide) throughout the study period. The in vitro antispasmodic activity on isolated tissues of rabbit jejunum on methanol extract showed concentration dependent (0.01-0.3 mg/ml) relaxation of spontaneous contractions with EC₅₀ value 0.011 µM and high K(+) (80 mM) induced contraction (0.01-0.1 mg/ml) with EC₅₀ value 0.066 mg/ml. Farsetia hamiltonii DCM and methanol extracts exhibited some antilipoxygenase activities while tyrosinase, acetylcholinesterase (AchE), carbonic II anhydrase, phosphodiesterase I, and antioxidant activity of plant extracts were not significant. Our results validate the traditional use of Farsetia hamiltonii for the traditional therapeutic potential in treating diabetes and gastrointestinal diseases.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Supelco
Methanol, analytical standard
Sigma-Aldrich
4-Nitrophenyl acetate, esterase substrate
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Baicalein, 98%
Sigma-Aldrich
Acarbose, ≥95% (HPLC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, Selectophore, ≥99.5%