Merck
  • Transcriptional characteristics associated with lichenysin biosynthesis in Bacillus licheniformis from Chinese Maotai-flavor liquor making.
Transcriptional characteristics associated with lichenysin biosynthesis in Bacillus licheniformis from Chinese Maotai-flavor liquor making.
Journal of agricultural and food chemistry (2015-01-07)
Qun Wu, Rong Zhang, Suqin Peng, Yan Xu
ABSTRACT
This work investigated the biosynthetic mechanism of lichenysin, the newly identified nonvolatile matrix component in Chinese liquors. Transcriptomes were analyzed in three producers, Bacillus licheniformis CGMCC 3961, 3962, and 3963, which were isolated from Maotai-flavor liquor-making process and produced 386.3, 553.5, and 795.2 μg/L lichenysin in a simulative liquor fermentation process. Lichenysin synthetase genes lchAA-AD in these three producers were expressed much more highly than those of the nonproducer B. licheniformis ATCC 14580 (>18.4-fold). In addition, ABC transporters were the most significant responsive metabolic pathway, and the expression levels of peptide transporter genes dppABCDE all increased more than 19.2-fold. When B. licheniformis CGMCC 3963 was cultured in synthetic medium, the expression of dppABCDE and lichenysin both increased with the addition of casein hydrolysate (containing various peptides). This indicated that peptide would act as a substrate for lichenysin synthesis. This work sheds new light on the mechanism for lichenysin biosynthesis.
Product Description
Supelco
Inositol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.99 atom % 12C
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 8x8mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%