Merck
  • The roles of integrins in function of human neutrophils after their migration through endothelium into interstitial matrix.
The roles of integrins in function of human neutrophils after their migration through endothelium into interstitial matrix.
PloS one (2015-02-24)
Ding Luo, Helen M McGettrick, Phil C Stone, George E Rainger, Gerard B Nash
ABSTRACT
We investigated the changes in neutrophil phenotype and function after transendothelial migration, and the roles played by integrin receptors in their behaviour. Neutrophils were tracked microscopically as they migrated through endothelial cells into collagen gels, and were retrieved at desired times. When endothelial cells were treated with increasing doses of tumour necrosis factor-α, neutrophils not only migrated in greater number, but also to a greater depth in the gel. Apoptosis was barely detectable in neutrophils retrieved after 24h, and many remained viable and motile at 48h. Neutrophils retrieved after 1h had increased oxidative capacity and at 24h had similar capacity as freshly-isolated neutrophils. However, by then they had impaired ability to phagocytose bacteria. Compared to fresh neutrophils, total mRNA was halved by 24h, but while β2-integrin expression decreased, β1- and β3-integrin increased along with ICAM-1. Studies of integrin blockade indicated that while β2-integrins were needed to cross the endothelial barrier, no integrins were required for migration within the gel. β2-integrins also contributed to phagocytosis, but their binding was not required for prolonged survival. These results demonstrate a model for integrated analysis of neutrophil migration and function, and describe development of effector functions and the roles of integrins in human neutrophils for the first time.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Arg-Gly-Asp-Ser, ≥95% (HPLC)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, purified by chromatography, ≥500 CDU/mg solid (CDU = collagen digestion units), lyophilized powder
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O