Merck
  • Norovirus infections and seroprevalence of genotype GII.4-specific antibodies in a Spanish population.
Norovirus infections and seroprevalence of genotype GII.4-specific antibodies in a Spanish population.
Journal of medical virology (2015-02-07)
Noelia Carmona-Vicente, Manuel Fernández-Jiménez, Juan M Ribes, Carlos J Téllez-Castillo, Parisá Khodayar-Pardo, Jesús Rodríguez-Diaz, Javier Buesa
ABSTRACT
Genotype II.4 noroviruses (NoVs) are a leading cause of epidemic acute gastroenteritis in children and adults worldwide. The prevalence of different NoV genotypes causing outbreaks and sporadic cases of acute gastroenteritis in the region of Valencia, Spain, during a 4-year period (2008-11) was investigated. NoVs were detected in 42 out of 55 (76.3%) outbreaks and in 26 out of 332 (7.8%) sporadic cases of acute gastroenteritis. Genogroup GII strains were predominant in outbreaks and sporadic cases. Different genotype GII.4 variants were found (Yerseke_2006a, Den Haag_2006b, Apeldoorn_2007, and New Orleans_2009), with the latter variant detected most frequently (35.3%). A recombinant P domain of the NoV GII.4 Apeldoorn_2007 variant was produced in Escherichia coli and used as the coating antigen in an enzyme immunoassay to survey the IgG antibody seroprevalence against NoV GII.4 in a Spanish population. Baculovirus-expressed virus-like particles (VLPs) of NoV GII.4 Den Haag_2006b variant were also produced and used as antigen to compare seroreactivity. Of the 434 serum specimens analyzed, 429 (98.6%) had antibodies against the P domain. The comparison of reactivities of 30 serum samples to the NoV GII.4 P polypeptide and VLP showed reproducible results with a correlation coefficient of r = 0.794. Titers of antibodies to the P domain increased gradually and significantly with age, reaching the highest levels at the age group of 41-50 years. These results confirm the high prevalence of NoV GII.4 infections in Valencia from early childhood.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Supelco
Urea, analytical standard
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Millipore
Urea solution, suitable for microbiology, 40% in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis