Merck
  • Characterization of antibody drug conjugate positional isomers at cysteine residues by peptide mapping LC-MS analysis.
Characterization of antibody drug conjugate positional isomers at cysteine residues by peptide mapping LC-MS analysis.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-18)
Marie-Claire Janin-Bussat, Marina Dillenbourg, Nathalie Corvaia, Alain Beck, Christine Klinguer-Hamour
ABSTRACT
Antibody-drug conjugates (ADCs) are becoming a major class of oncology therapeutics. Because ADCs combine the monoclonal antibody specificity with the high toxicity of a drug, they can selectively kill tumor cells while minimizing toxicity to normal cells. Most of the current ADCs in clinical trials are controlled, but heterogeneous mixtures of isomers and isoforms. Very few protocols on ADC characterization at the peptide level have been published to date. Here, we report on the improvement of an ADC peptide mapping protocol to characterize the drug-loaded peptides by LC-MS analysis. These methods were developed on brentuximab vedotin (Adcetris), a commercial ADC with an average of four drugs linked to interchain cysteine residues of its antibody component. Because of the drug hydrophobicity, all the steps of this protocol including enzymatic digestion were improved to maintain the hydrophobic drug-loaded peptides in solution, allowing their unambiguous identification by LC-MS. For the first time, the payloads positional isomers observed by RP-HPLC after IdeS-digestion and reduction of the ADC were also characterized.
Product Description
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Millipore
Bifido Selective Supplement B, suitable for microbiology
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%