Merck
  • Characterizing metabolites and potential metabolic pathways for the novel psychoactive substance methoxetamine.
Characterizing metabolites and potential metabolic pathways for the novel psychoactive substance methoxetamine.
Drug testing and analysis (2014-02-28)
E L Menzies, S C Hudson, P I Dargan, M C Parkin, D M Wood, A T Kicman
ABSTRACT
The classic approach of controlled volunteer studies to study drug metabolism is difficult or impossible to undertake for novel psychoactive substances (NPS), as there is generally very limited information available to allow appropriate dose finding and safety. A viable and powerful alternative is the identification and characterization of phase I and II metabolites of such drugs by examining the concordance of data gathered from analysis of microsomal incubates with that from analysis of specimens collected from individuals with analytically confirmed use of NPS. Liquid chromatography-high resolution mass spectrometry provides the ability to reliably identify such metabolites. We used this technique here to study the metabolism of the ketamine analogue methoxetamine. A large number of metabolites were identified in the in vitro studies including normethoxetamine, O-desmethylmethoxetamine, dihydromethoxetamine, dehydromethoxetamine and several structural isomers of hydroxymethoxetamine and hydroxynormethoxetamine. pH dependent liquid-liquid extraction was used to discriminate phenolic from alcoholic metabolites. Phase II glucuronide conjugates included those of O-desmethylmethoxetamine, O-desmethylnormethoxetamine and O-desmethylhydroxymethoxetamine. The majority of these phase I and II metabolites were confirmed to be present in urine collected from three individuals presenting with acute methoxetamine toxicity.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioXtra, ≥98.0% (T)
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
1-Chlorobutane, ReagentPlus®, 99%
Sigma-Aldrich
1-Chlorobutane, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Sodium phosphate monobasic dihydrate, BioUltra, Molecular Biology, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic dihydrate, purum p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium phosphate monobasic dihydrate, meets analytical specification of Ph. Eur., BP, USP, E 339, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioUltra, Molecular Biology, ≥99.0% (T)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets