Merck
  • Integrated microfluidic chip and online SCX separation allows untargeted nanoscale metabolomic and peptidomic profiling.
Integrated microfluidic chip and online SCX separation allows untargeted nanoscale metabolomic and peptidomic profiling.
Journal of proteome research (2015-01-13)
Ravi Tharakan, Dingyin Tao, Ceereena Ubaida-Mohien, Rhoel R Dinglasan, David R Graham
ABSTRACT
Metabolomics and peptidomics are systems biology approaches in which broad populations of molecular species produced in a cell or tissue sample are identified and quantified. These two molecular populations, metabolites and peptides, can be extracted from tissues in a similar fashion, and we therefore have here developed an integrated platform for their extraction and characterization. This was accomplished by liquid-liquid extraction of peptides and metabolites from tissue samples and online strong cation exchange (SCX) separation to allow characterization of each population individually. The platform was validated both by a mixed set of purified standards and by an analysis of splenic tissue from SIV-infected macaques, showing both good reproducibility in chromatography, with relative standard deviation (RSD) of hold time less than 0.4%, and clear separation of charge state, with ∼ 95% of molecular features in SCX separated runs at charge states of +1 or +2. Finally, we used this platform to analyze the physiological response to infection in the spleen, showing that the spleen contains an abundance of hemoglobin-derived peptides, which do not appear to change in response to infection, and that there appears to be a large and variable metabolic response to infection. We therefore present a method for peptidomic and metabolomic profiling which is simple, robust, and easy to implement.
Product Description
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ultrapure Acetonitrile
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)