Merck
Comparison of UPLC and HPLC methods for determination of vitamin C.
Food chemistry (2015-01-13)
Inga Klimczak, Anna Gliszczyńska-Świgło
ABSTRACT
Ultra performance liquid chromatography (UPLC) and high-performance liquid chromatography (HPLC) methods for determination of ascorbic acid (AA) and total AA (TAA) contents (as the sum of AA and dehydroascorbic acid (DHAA) after its reduction to AA) in fruit beverages and in pharmaceutical preparations were compared. Both methods are rapid: total time of analysis was 15 and 6 min for HPLC and UPLC methods, respectively. The methods were validated in terms of linearity, instrument precision, limits of detection (LOD) and quantification (LOQ), accuracy and recovery. Intra- and inter-day instrument precisions for fruit juices, expressed as RSD, were 2.2% and 2.4% for HPLC, respectively, and 1.7% and 1.9% for UPLC, respectively. For vitamin C tablets, inter- and intra-day precisions were 0.4% and 0.5%, respectively (HPLC), and 0.5% and 0.3%, respectively (UPLC). Both methods were sensitive: LOD was 0.049 μg/mL for HPLC and 0.024 μg/mL for UPLC while LOQs were 0.149 and 0.073 μg/mL for HPLC and UPLC, respectively. These methods could be useful in the routine qualitative and quantitative analysis of AA or TAA in pharmaceutical preparations or fruit beverages. However, UPLC method is more sensitive, faster and consumes less eluent.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
DL-Dithiothreitol, suitable for electrophoresis, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, Molecular Biology, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
L-Ascorbic acid, analytical standard
Supelco
Methanol, analytical standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%