Merck
  • Anti-trypanosomal activity of diarylheptanoids isolated from the bark of Alnus japonica.
Anti-trypanosomal activity of diarylheptanoids isolated from the bark of Alnus japonica.
The American journal of Chinese medicine (2014-09-03)
Nguyen Huu Tung, Mitsuko Suzuki, Takuhiro Uto, Osamu Morinaga, Kofi D Kwofie, Naa Ammah, Kwadwo A Koram, Frederic Aboagye, Dominic Edoh, Taizo Yamashita, Yasuchika Yamaguchi, Takao Setsu, Shoji Yamaoka, Nobuo Ohta, Yukihiro Shoyama
ABSTRACT
The crude extract of Alnus japonica bark exhibited a strong effect on the growth of Trypanosoma brucei. Subsequent chromatographic separation resulted in the isolation of two novel diarylheptanoids, known as alnuside C (2) and alnuside D (3), and three known compounds, 1-(3,4-dihydroxyphenyl)-7-(4-hydroxyphenyl)-heptan-3(R)-O-β-D-glucopyranoside (1), oregonin (4) and hirsutanone (5). The structures of the isolates were elucidated based on the use of extensive spectroscopic and chemical methods. Among the isolated diarylheptanoids, oregonin (4) (a major component of plant bark) and hirsutanone (5) exhibited potent in vitro inhibitory activity against T. brucei growth in the bloodstream with IC50 values of 1.14 and 1.78 μM, respectively. We confirmed that oregonin (4) and hirsutanone (5) were not toxic to human normal skin fibroblast cells (NB1RGB) and colon cancer cells (HCT-15) at a concentration of 50 μM; however, lower levels of toxicity were observed for leukemia cells. To determine the structure activity relationships of the isolated components, we performed Conformation Search and found that the 3-oxo function of the heptane chain in the diarylheptanoid molecule is required for their trypanocidal activity.
Product Description
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%