Merck
  • Caffeine induces cardiomyocyte hypertrophy via p300 and CaMKII pathways.
Caffeine induces cardiomyocyte hypertrophy via p300 and CaMKII pathways.
Chemico-biological interactions (2014-08-06)
Liang Shi, Hao Xu, Jinhong Wei, Xingfeng Ma, Jianbao Zhang
ABSTRACT
Caffeine is commonly utilized to trigger intracellular calcium in cardiomyocyte. It is well accepted that caffeine could induce cardiac arrhythmia, but it is not clear with regard of its impacts on the cardiac function. This article presents a recent study concerning the effects of caffeine on the cardiomyocyte hypertrophy and the associated signal pathway. The experimental results showed that the total protein contents, the surface area of cardiomyocyte and β-myosin heavy chain (β-MHC) expression increased in ventricular myocytes of neonatal Sprague-Dawley (SD) rats after 24h caffeine incubation. It is also observed that the basal intracellular calcium (Ca(2+)) level has increased, while the amplitude of Ca(2+) oscillation and Ca(2+) content have decreased in sarcoplasmic reticulum (SR). The caffeine-induced myocyte enhancer factor-2 (MEF2) expression and hypertrophy can be completely abolished by the inhibition of cardiac ryanodine receptor (RyR2), as well as KN93 and curcumin treatments. Meanwhile, the amplitude of Ca(2+) oscillation and the Ca(2+) content of SR in the completely-inhibited group have reached the physiological level. These results suggest that the caffeine-induced cardiomyocyte hypertrophy established the connection between Ca(2+) release from SR and cytosol that activates CaMKII and p300, which in turn enhances the expression of MEF2 that promotes cardiomyocyte hypertrophy.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O